WHA10408712
Whatman® mixed cellulose ester ME 24 filter discs 0.2 μm pore size
white, hydrophilic, black grid (3.1 mm), sterile, for membrane butler, 100 EA, 400 ea, 47 mm diam
Synonym(s):
Z750476
About This Item
Recommended Products
product name
Whatman® ME membranes for microbiological control, ME24/21, without interleaving paper, pore size 0.2 μm, black grid, diam. 50 mm
material
black grid
packaging
pk of 100
manufacturer/tradename
Whatman 10408712
Whatman Article No. 28415241 (US reference)
diam.
50 mm
membrane diam.
47 mm
pore size
0.2 μm
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
- Excellent contrast for easy particle detection
- High internal surface area for sample adsorption
- High dirt loading capacity
- Biologically inert with good thermal stability
- Uniform microporous structure delivers high flow rates
Other Notes
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service