Skip to Content
Merck
All Photos(1)

Documents

W522406

Sigma-Aldrich

Cedarwood oil, Texas

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Juniperus mexicana Schiede

grade

Fragrance grade

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

origin

USA origin

refractive index

n20/D 1.505 (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

woody

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Extraction method: steam distillation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaun Yon-Seng Khoo et al.
Frontiers in behavioral neuroscience, 14, 5-5 (2020-03-03)
Conditioned responding can be renewed by re-exposure to the conditioning context following extinction in a different context (ABA renewal) or by removal from the extinction context (AAB or ABC renewal). ABA renewal is robust in Pavlovian and operant conditioning paradigms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service