Skip to Content
Merck
All Photos(1)

Key Documents

W358800

Sigma-Aldrich

1,3-Propanedithiol

≥96%, FG

Synonym(s):

1,3-Dimercaptopropane, Trimethylene dimercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HS(CH2)3SH
CAS Number:
Molecular Weight:
108.23
FEMA Number:
3588
Beilstein:
1071197
EC Number:
Council of Europe no.:
11929
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.076
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
To inquire about this product W358800, please contact your local Merck office or dealer. Contact Technical Service

biological source

synthetic

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

vapor density

>1 (vs air)

vapor pressure

5 mmHg ( 37.7 °C)

Assay

≥96%

refractive index

n20/D 1.539 (lit.)

bp

169 °C (lit.)

mp

−79 °C (lit.)

density

1.078 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty; sulfurous

SMILES string

SCCCS

InChI

1S/C3H8S2/c4-2-1-3-5/h4-5H,1-3H2

InChI key

ZJLMKPKYJBQJNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service