Skip to Content
Merck
All Photos(1)

Key Documents

W344818

Sigma-Aldrich

Dicyclohexyl disulfide

≥95%, FG

Synonym(s):

Cyclohexyl disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11S-)2
CAS Number:
Molecular Weight:
230.43
FEMA Number:
3448
EC Number:
Council of Europe no.:
2320
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.028
NACRES:
NA.21
Organoleptic:
onion; alliaceous
grade:
FG
Halal
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

FG
Halal

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

≥95%

refractive index

n20/D 1.545 (lit.)

bp

162-163 °C/6 mmHg (lit.)

density

1.046 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

onion; alliaceous

SMILES string

C1CCC(CC1)SSC2CCCCC2

InChI

1S/C12H22S2/c1-3-7-11(8-4-1)13-14-12-9-5-2-6-10-12/h11-12H,1-10H2

InChI key

ODHAQPXNQDBHSH-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service