W289906
3-Phenylpropyl isovalerate
≥99%
Synonym(s):
3-Phenylpropyl-β-methylbutyrate, Hydrocinnamyl isovalerate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)2CHCH2CO2(CH2)3C6H5
CAS Number:
Molecular Weight:
220.31
FEMA Number:
2899
EC Number:
Council of Europe no.:
462
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
biological source
synthetic
Assay
≥99%
refractive index
n20/D 1.484 (lit.)
bp
285 °C (lit.)
density
0.98 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
SMILES string
CC(C)CC(OCCCC1=CC=CC=C1)=O
InChI
1S/C14H20O2/c1-12(2)11-14(15)16-10-6-9-13-7-4-3-5-8-13/h3-5,7-8,12H,6,9-11H2,1-2H3
InChI key
LBNFCOMOXOWXCD-UHFFFAOYSA-N
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service