Skip to Content
Merck
All Photos(1)

Documents

W218006

Sigma-Aldrich

Isobutyl angelate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=C(CH3)CO2CH2CH(CH3)2
CAS Number:
Molecular Weight:
156.22
FEMA Number:
2180
EC Number:
Council of Europe no.:
247
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.408

availability

available only in EU

refractive index

n20/D 1.434 (lit.)

bp

176-177 °C (lit.)

density

0.877 g/mL at 25 °C (lit.)

SMILES string

C\C=C(\C)C(=O)OCC(C)C

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service