Skip to Content
Merck
All Photos(1)

Key Documents

W216402

Sigma-Aldrich

Bornyl valerate

≥96%, mixture of isomers

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26O2
CAS Number:
Molecular Weight:
238.37
FEMA Number:
2164
EC Number:
Council of Europe no.:
471
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.153

Assay

≥96%

optical activity

[α]20/D −18°, neat

impurities

15-30% isobornyl valerate

refractive index

n20/D 1.463 (lit.)

bp

136 °C/12 mmHg (lit.)

density

0.952 g/mL at 25 °C (lit.)

SMILES string

CCCCC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C

Biochem/physiol Actions

Taste at 1.0 ppm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 24, 49-49 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service