Skip to Content
Merck
All Photos(1)

Documents

QBD10540

Sigma-Aldrich

m-dPEG ®24-Azide (Azido-m-dPEG ®24)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H99N3O24
CAS Number:
Molecular Weight:
1114.32
MDL number:
UNSPSC Code:
12352125
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: chemical modification reagent
reagent type: cross-linking reagent
reaction type: click chemistry

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

m-dPEG ®₂₄-Azide (Azido-m-dPEG ®₂₄) can be used for adding a hydrophilic dPEG modifier by copper-catalyzed "click" chemistry in which the azide is used to react with terminal alkynes or strained alkynes.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service