Key Documents
PL800GA-NIT-9
Plas-Labs replacement nitrile gloves
for 2 piece glove set, size 9 (for 8 in. ports)
Select a Size
1 PAIR
€114.00
€114.00
Please contact Customer Service for Availability
Select a Size
Change View
1 PAIR
€114.00
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
€114.00
Please contact Customer Service for Availability
material
nitrile
size
9 (for 8 in. ports)
feature
reusable
for use with glove box sleeves
packaging
pack of 1 pair
manufacturer/tradename
Plas-Labs 800GA-NIT-9
General description
Plas-Labs glovebox gloves are available as either one-piece or two-piece assemblies with interchangeable sleeves and gloves. This enables personnel to use the same glove box by changing the size or type of glove being used. Choose from four rubber types. Gloves are 15 mil thickness unless noted otherwise.
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
  • White Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light
  • Black neoprene rubber for chemical resistance
  • Black butyl rubber for highest impermeability to water vapor, gases, and toxic fluids
  • Green nitrile rubber for resistance to oil, fuel, and acids
  • Latex rubber for flexibility
Legal Information
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)