Skip to Content
Merck
All Photos(1)

Key Documents

MT51302620

Mettler-Toledo SevenGo Duo Pro pH/Ion/conductivity meter SG78

SG478 BASIC, no electrode

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115603
NACRES:
NB.61
Product MT51302620 is not currently sold in your country. Contact Technical Service

description

Calibration pH: Max. 5 points, 7 predefined buffer groups
Conductivity range 0.01 μ (+/- 0.5%)
Connection to computer IR interface for printer or PC (via RS232 or USB)
Resistivity range 0.01 … 100.00 MOhm•cm

manufacturer/tradename

Mettler-Toledo 51302620

TDS measuring range

0.1 mg/L (min)
600 g/L (max)

mV measuring range

-1999.9-1999.9, accuracy: ±0.1

salinity measuring range

0-80 (psu)

temperature measuring range

-5-105 °C, accuracy: ±0.2

battery power

4 AA (included in standard delivery)

pH range

-2-19.999

Looking for similar products? Visit Product Comparison Guide

General description

Includes meter only.
SevenGo Duo pro combines maximum measurement accuracy and an excellent range of functions with user-friendly operation and exceptional security. SevenGo Duo pro meters are essential for special analyses, particularly in regulated sectors in the pharmaceutical, chemical, cosmetic and food & beverage industry. The kit versions are also well suited for outdoor use because both the meter and the sensors are watertight (IP67).

The revolutionary intelligent sensor management system (ISM®) offers the user an even greater level of security. The calibration data is stored on the chip in the sensor and is used automatically when the sensor is connected to the meter. This ensures that the meter always uses the latest calibration data.

Features and Benefits
pH/Ion/Conductivity Meter (IP67) for the Strictest Demands

Portable dual-channel meter for the parallel determination of pH, mV, rel. mV or ion concentrations and conductivity, TDS (total dissolved solids), salinity or specific resistance

Either linear or segmented 5-point pH calibration with selection from 7 predefined and one user-defined buffer groups

5 predefined conductivity standards – option to enter user-defined standard or cell constant
Modern data management for 500 GLP data records with date/time, sensor ID and serial number, user and sample ID

Contactless infrared interface for data transmission to a printer or PC
Intuitive, user-friendly operation using menu guidance in 10 languages and backlit display

Rugged watertight housing (IP67)

Revolutionary intelligent sensor management (ISM®)

Legal Information

ISM is a registered trademark of Mettler-Toledo, Inc.
SevenGo is a trademark of Mettler-Toledo, Inc.
SevenGo Duo pro is a trademark of Mettler-Toledo, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service