Skip to Content
Merck
All Photos(1)

Key Documents

L060002-HH

Sigma-Aldrich

Oxidizer 0.02M

Synonym(s):

Oxidizer

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116105

type

for DNA synthesis

product line

Proligo Reagents

packaging

pkg of 4 × 2.5 L (L060252-04)

concentration

5.4-6.6 g/L iodine (titration)

color

clear brownish, solution

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Tetrahyrofurane/Water/Pyridine/Iodine 66/12/22/0.6 (v/v/v/w)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 3

Target Organs

Central nervous system, Respiratory system, Thyroid

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

<50.0 °F - closed cup

Flash Point(C)

< 10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service