GF31603115
Polymethylpentene
TPX®, granule, 3 mm nominal granule size, weight 1000 g, grade dx560m
Synonym(s):
ME316306
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
granular
manufacturer/tradename
Goodfellow 316-031-15
weight
1000 g
nominal granule size
3 mm
Related Categories
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
TPX is a registered trademark of Mitsui Petrochemical Industries, Ltd.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service