DWK986724
WHEATON® liquid scintillation vial with seperate unlined PE cap
transparent high-density polyethylene bottle, capacity (20 mL), screw cap, case of 1,000 ea Bulk packed vials with screw caps in separate bag
Synonym(s):
986724
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
transparent high-density polyethylene bottle
white polyethylene cap
feature
with cap
packaging
case of 1,000 ea Bulk packed vials with screw caps in separate bag
manufacturer/tradename
WHEATON® 986724
bottle diam. × H × W
(27 mm) (61 mm) (375 mm)
capacity
(20 mL)
fitting
22-400 neck
22-400
Looking for similar products? Visit Product Comparison Guide
General description
Liquid Scintillation Vials with separately packs caps
- For optimal performance in the laboratory, PET plastic vials provide the clarity of glass and have the safety of using plastic.
- Vials can be safely incinerated; no harmful gas is generated; only CO2 and H2O are created.
- Lightweight, and virtually unbreakable PET vials offer low permeability to solvents and minimal background counts.
- 20 mL size
- Made from polyethylene terephthalate (PET)
- Clarity of glass with safety of plastic
- Low permeability to solvents and minimal background count
- Dimensions (OD x H): 27 mm x 61 mm (with cap attached)
Features and Benefits
• Converted from Type I borosilicate glass tubing, PET, or HDPE• Background counts are consistent and low; ultraviolet transmission is high
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
related product
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service