CLS55640100
Pyrex® class A low-actinic volumetric flask, Pyrex® ST stopper
capacity 100 mL
Synonym(s):
flasks glass, volumetric flasks, volumetric glass
About This Item
Recommended Products
material
borosilicate glass
long
Joint
joint stopper13
sterility
non-sterile
feature
class A
closure type ST stopper
packaging
case of 12
pack of 6
manufacturer/tradename
Corning 55640-100
Corning 55640
tol. measuring range
0.08 mL
capacity
100 mL
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service