CLS4442250
Pyrex® long neck culture flask
capacity 250 mL
Synonym(s):
conical flasks, Corning® 4442
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
Delong
Delong neck
borosilicate glass
flat bottom flask
sterility
non-sterile
feature
cap: no
closure type closure cap
packaging
case of 12
pack of 6
manufacturer/tradename
Corning 4442-250
capacity
250 mL
neck O.D.
(38 mm)
38 mm
Looking for similar products? Visit Product Comparison Guide
General description
Neck is made from heavy-wall, rimless tubing and reduces the amount of splashed solution reaching the closure. Ideal for shaker cultures.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service