Key Documents
CLS3540
Corning® 384 well microplate, low volume
black polystyrene, round bottom, non-sterile, lid: no, pack of 10
Synonym(s):
384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates
Select a Size
50 EA
€705.00
€705.00
Please contact Customer Service for Availability
Select a Size
Change View
50 EA
€705.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.24
material:
black polystyrene
clear bottom
clear wells
round bottom
size:
384 well
sterility:
non-sterile
binding type:
non-treated surface
feature:
lid: no
skirt
€705.00
Please contact Customer Service for Availability
material
black polystyrene
clear bottom
clear wells
round bottom
sterility
non-sterile
feature
lid: no
skirt
packaging
pack of 10
case of 50
manufacturer/tradename
Corning 3540
size
384 well
well volume
50 μL
well working volume
5-40 μL
wells
384 wells , low volume
suitability
suitable for (molecular, validated for the FLIPR reader)
binding type
non-treated surface
General description
  • Corning® 384 well low volume microplates feature a round well design with a clear flat bottom.
  • The non-treated medium binding polystyrene surface is hydrophobic and suitable for the immobilization of antibodies.
  • Ideal for fluorescent homogeneous assays.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)