Skip to Content
Merck
All Photos(1)

Key Documents

CDS022536

Sigma-Aldrich

Lenalidomide

Synonym(s):

3-(7-Amino-3-oxo-1H-isoindol-2-yl)piperidine-2,6-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13N3O3
Molecular Weight:
259.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

description

AldrichCPR

form

powder

SMILES string

O=C1N(C2CCC(NC2=O)=O)CC3=C1C=CC=C3N

InChI

1S/C13H13N3O3/c14-9-3-1-2-7-8(9)6-16(13(7)19)10-4-5-11(17)15-12(10)18/h1-3,10H,4-6,14H2,(H,15,17,18)

InChI key

GOTYRUGSSMKFNF-UHFFFAOYSA-N

Gene Information

Application

Lenalidomide is a structural analog of thalidomide, which exhibits excellent anticancer activity against multiple myeloma.[1][2] It can be used in synthesizing multiple derivatives of lenalidomide to study their biological activity.[3] Synthesis of gelator containing lenalidomide, which forms stable gels in 1,4-dioxane has been reported.[4]

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2

Target Organs

Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lenalidomide plus dexamethasone for relapsed or refractory multiple myeloma.
Dimopoulos M, et al.
The New England Journal of Medicine, 357(21), 2123-2132 (2007)
Melamine tunable effect in a lenalidomide-based supramolecular self-assembly system via hydrogen bonding.
Gao A, et al.
New. J. Chem., 41(16), 7924-7931 (2017)
Cereblon is a direct protein target for immunomodulatory and antiproliferative activities of lenalidomide and pomalidomide.
Lopez-Girona A E A, et al.
Leukemia, 26(11), 2326-2326 (2012)
Design, synthesis and biological evaluation of Lenalidomide derivatives as tumor angiogenesis inhibitor.
Hu S, et al.
Bioorganic & Medicinal Chemistry Letters, 27(17), 4075-4081 (2017)
Xiaofei He et al.
Experimental hematology, 86, 21-27 (2020-05-22)
Our previous study revealed that expression of G protein-coupled receptor 68 (GPR68) was upregulated in MDSL cells, a cell line representing myelodysplastic syndromes (MDS), in response to lenalidomide (LEN), and mediated a calcium/calpain proapoptotic pathway. Isx, a GPR68 agonist, enhanced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service