Skip to Content
Merck
All Photos(1)

Key Documents

CBR00181

Sigma-Aldrich

4-Quinazolinamine

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N3
CAS Number:
Molecular Weight:
145.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

NC1=C(C=CC=C2)C2=NC=N1

InChI

1S/C8H7N3/c9-8-6-3-1-2-4-7(6)10-5-11-8/h1-5H,(H2,9,10,11)

InChI key

DRYRBWIFRVMRPV-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Legal Information

Product of ChemBridge Corp.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Moyal et al.
The British journal of dermatology, 177(3), 791-800 (2017-03-04)
MicroRNA (miR)-155 contributes to the proliferation of mycosis fungoides (MF) in vitro and is upregulated in tumours of MF compared with early MF lesions. To investigate the contribution of miR-155 to the cancerous phenotype and drug resistance of MF/Sézary cell
Clélia Dental et al.
Journal of immunology (Baltimore, Md. : 1950), 198(3), 1229-1241 (2016-12-21)
A shock-and-kill approach involving the simultaneous treatment of HIV-1-infected patients with latency-reversing agents (LRAs) and combination antiretroviral therapy was proposed as a means to eradicate viral reservoirs. Currently available LRAs cannot discriminate between HIV-1-infected and uninfected cells. Therefore, the risks
Minghui Liu et al.
Oncotarget, 7(51), 84951-84964 (2016-11-09)
Acquired resistance to epidermal growth factor receptor tyrosine kinase inhibitors (EGFR-TKIs) is a major challenge to targeted therapy for non-small cell lung cancer (NSCLC). We investigated whether a cyclin D kinase 4/6 (CDK4/6) inhibitor, PD 0332991, could reverse EGFR-TKI resistance
Ilaria Cristofaro et al.
Cells, 9(3) (2020-03-19)
Glioblastomas (GBM) are the most aggressive form of primary brain tumors in humans. A key feature of malignant gliomas is their cellular heterogeneity. In particular, the presence of an undifferentiated cell population of defined Glioblastoma Stem cells (GSCs) was reported.
Kota Naito et al.
Biological & pharmaceutical bulletin, 42(4), 631-637 (2019-02-05)
Serotonin (5-hydroxytryptamine; 5-HT) can induce hepatocyte DNA synthesis and proliferation by autocrine secretion of transforming growth factor (TGF)-α through 5-HT2B receptor/phospholipase C (PLC)/Ca2+ and a signaling pathway involving epidermal growth factor (EGF)/TGF-α receptor tyrosine kinase (RTK)/extracellular signal-regulated kinase 2 (ERK2)/mammalian

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service