CAD7953
Laboratory pipetting needle with 90° blunt ends
gauge 17, L 4 in., 304 stainless steel hub
All Photos(1)
About This Item
Recommended Products
material
304 stainless steel hub
packaging
pack of 12 ea
manufacturer/tradename
Cadence Science 7953
L
4 in.
gauge
17
Related Categories
General description
Pipetting needles are available in a wide choice of sizes for use in laboratory procedures. All needles are made with type 304 stainless steel cannula.
Catalog numbers CAD7950 to CAD7957 needles are fitted with stainless steel hubs for special applications with corrosive materials or critical procedures. Catalog numbers CAD7930 to CAD7942 needles are fitted with standard nickel plated Luer hubs. These needles are used in conjunction with both manual and automatic volumetric dispensers, dilutors, and pipettors.
Catalog numbers CAD7950 to CAD7957 needles are fitted with stainless steel hubs for special applications with corrosive materials or critical procedures. Catalog numbers CAD7930 to CAD7942 needles are fitted with standard nickel plated Luer hubs. These needles are used in conjunction with both manual and automatic volumetric dispensers, dilutors, and pipettors.