Skip to Content
Merck
All Photos(1)

Documents

C64207

Sigma-Aldrich

4-Chlorophenyl cyclopropyl ketone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4COC3H5
CAS Number:
Molecular Weight:
180.63
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.5718 (lit.)

mp

29-31 °C (lit.)

SMILES string

Clc1ccc(cc1)C(=O)C2CC2

InChI

1S/C10H9ClO/c11-9-5-3-8(4-6-9)10(12)7-1-2-7/h3-7H,1-2H2

InChI key

OPSFCTBBDIDFJM-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service