Skip to Content
Merck
All Photos(1)

Documents

BMSBT4500

Benchmark Orbi-Shaker CO2 with remote controller

AC/DC input 115 V

Synonym(s):

orbital shaker, microplate shaker, carbon dioxide incubator, CO2 incubation, harsh enviroment incubation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103811
NACRES:
NB.73

manufacturer/tradename

Benchmark Scientific BT4500

parameter

200-1200 rpm speed

plate W × L

33 cm × 30 cm

Looking for similar products? Visit Product Comparison Guide

General description

OrbiShaker CO2MP is designed for thorough, mixing of microplates in extreme environments, including CO2 incubators. It can withstand temperatures from 0 to 65°C and humidity levels up to 100%.

Application

Accepts 6 microplates, designed for use in CO2 incubators

Legal Information

Orbi-Shaker is a trademark of Benchmark Scientific, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service