Skip to Content
Merck
All Photos(1)

Key Documents

B4201

Sigma-Aldrich

1,2,3-Benzenetricarboxylic acid hydrate

98%

Synonym(s):

Hemimellitic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(CO2H)3·xH2O
CAS Number:
Molecular Weight:
210.14 (anhydrous basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

crystals

mp

190-192 °C (dec.) (lit.)

SMILES string

O.OC(=O)c1cccc(C(O)=O)c1C(O)=O

InChI

1S/C9H6O6.H2O/c10-7(11)4-2-1-3-5(8(12)13)6(4)9(14)15;/h1-3H,(H,10,11)(H,12,13)(H,14,15);1H2

InChI key

DYDNZUYNRZENMU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olga Catalina-Rodriguez et al.
Oncotarget, 3(10), 1220-1235 (2012-10-27)
Dysregulation of the pathways that preserve mitochondrial integrity hallmarks many human diseases including diabetes, neurodegeration, aging and cancer. The mitochondrial citrate transporter gene, SLC25A1 or CIC, maps on chromosome 22q11.21, a region amplified in some tumors and deleted in developmental
Kevin H Lin et al.
Cell metabolism, 29(5), 1217-1231 (2019-02-19)
Crosstalk between metabolic and survival pathways is critical for cellular homeostasis, but the connectivity between these processes remains poorly defined. We used loss-of-function CRISPR/Cas9 knockout screening to identify metabolic genes capable of influencing cellular commitment to apoptosis, using sensitization to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service