Skip to Content
Merck
All Photos(2)

Key Documents

931195

Sigma-Aldrich

CX401

Umicore

Synonym(s):

Methanesulfonato(tris(1-adamantyl)phosphine)[2-(2′-amino-1,1′- biphenyl)]palladium(II), Pd(PAd3)G3, [Pd(PAd3)[2-(2′-amino-1,1′-biphenyl)](Ms)]

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€136.00
1 G
€295.00

€136.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€136.00
1 G
€295.00

About This Item

Empirical Formula (Hill Notation):
C43H58NO3PPdS
CAS Number:
Molecular Weight:
806.38
MDL number:
UNSPSC Code:
12352142
NACRES:
NA.21

€136.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

powder

mp

208 °C (decomposition)

storage temp.

2-8°C

Application

CX401 is the 3rd generation Pd precatalyst of P(Ad3).

Storage and Stability

Do not store above 25 °C (77 °F). Store in a tightly closed original container under dry inert gas, protected from direct sunlight in a dry, cool and well-ventilated area.

Legal Information

Product of Umicore.

This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

Additional information available at www.pmc.umicore.com

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service