Skip to Content
Merck
All Photos(3)

Documents

929697

Sigma-Aldrich

ElectroGreen®

greener alternative

Butyl Cellosolve substitute for electronics, bio-sourced

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000
NACRES:
NA.05

grade

for electronic purposes

Quality Level

vapor pressure

1.75 mmHg ( 20 °C)

description

RelativeEvaporation rate: ~0.11

Hansen SolubilityParameters: SPd = 7.5; SPp = 3.8; SPh = 6.1

Assay

≥99% (GC)

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

impurities

≤0.2% Water (Karl Fischer)
≤1 ppm As, Cr, Cd, Cu, Hg, Mn, Ni, Pb, Zn, trace (ICP, each)

evapn. residue

≤0.05%

color

clear

viscosity

16.1  cP(20 °C)

bp

154 °C (Initial)

density

0.954

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

This ElectroGreen® solvent is made of bio-based (corn/beets derived) safer alternatives to replace synthetic butyl cellosolve fossil-based solvent with specifications tailored for electronics and energy applications. All our ElectroGreen® solvents line are derived from a renewable resource(verified through C14 ASTM D6866-16 testing). We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Click here for more information.

This solvent blend consists of Ethyl lactate 70-90%, hexan-1-ol 30-50%.

Application

Our bio-based replacement for Butyl Cellosolve is perfect for cleaning in electronics, as well as ink formulations.
It is a precision cleaning solvent, dries completely, no film residue. It is also easy and inexpensive to distill/recycle, whilst no environmentally hazardous ingredients and no Hazardous Air Pollutants.
It is 20% to 30% more efficient in viscosity reduction than Butyl Cellosolve. It is an excellent solvency for ink formulations with high loading capacity.

Legal Information

ElectroGreen is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

141.0 °F

Flash Point(C)

60.55 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Carbon-based Sustainable Organic Electronics (SOE) limit the use of critical elements and biodegrade at their end-of-life. This review offers insight on how structural and energy disorder in these materials influence device performance and includes evaluations of various transport models and their limitations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service