Skip to Content
Merck
All Photos(1)

Key Documents

926191

Sigma-Aldrich

Ru(OAc)2[(R)-dtbm-SEGPHOS®]

Takasago

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€52.10
1 G
€152.00
5 G
€365.00

€52.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€52.10
1 G
€152.00
5 G
€365.00

About This Item

Empirical Formula (Hill Notation):
C78H106O12P2Ru
CAS Number:
Molecular Weight:
1398.69
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

€52.10


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

powder

Application

Ru(OAc)2[(S)-dtbm-SEGPHOS®] is a ruthenium catalyst for asymmetric hydrogenation of olefins.

Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Legal Information

SEGPHOS is a registered trademark of Takasago Intl. Corp.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel J Tindall et al.
Angewandte Chemie (International ed. in English), 60(2), 721-725 (2020-09-15)
Sugar alcohols are of great importance for the food industry and are promising building blocks for bio-based polymers. Industrially, they are produced by heterogeneous hydrogenation of sugars with H2 , usually with none to low stereoselectivities. Now, we present a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service