Skip to Content
Merck
All Photos(4)

Key Documents

924628

Sigma-Aldrich

Cross-shaped channel chip - 50 μm channel (extended size platform ΙΙ)

Fluidic 189, Cyclic Olefin Copolymer (COC) with Luer Interface

Synonym(s):

Microfluidic

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€162.00

€162.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
€162.00

About This Item

UNSPSC Code:
42142600
NACRES:
NA.23

€162.00


Please contact Customer Service for Availability

Request a Bulk Order

Application

This is a Cross-shaped channel chips with Extended size platform ΙΙ with a Luer fluidic interface.

Cross-shaped channel chip - 50 μm channel with extended size platform II, Fluidic 189, Cyclic Olefin Copolymer COC) with Luer Interface.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent advances of controlled drug delivery using microfluidic platforms.
Sanjay, et al.
Advanced Drug Delivery Reviews, 128, 3-28 (2020)
Dongfei Liu et al.
Lab on a chip, 17(11), 1856-1883 (2017-05-10)
The microfluidic technique has brought unique opportunities toward the full control over the production processes for drug delivery carriers, owing to the miniaturisation of the fluidic environment. In comparison to the conventional batch methods, the microfluidic setup provides a range
Sharma T Sanjay et al.
Advanced drug delivery reviews, 128, 3-28 (2017-09-19)
Conventional systematically-administered drugs distribute evenly throughout the body, get degraded and excreted rapidly while crossing many biological barriers, leaving minimum amounts of the drugs at pathological sites. Controlled drug delivery aims to deliver drugs to the target sites at desired

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service