Skip to Content
Merck
All Photos(1)

Key Documents

908371

Methyl (Z)-2-phenyl-3-(tosyloxy)acrylate

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16O5S
CAS Number:
Molecular Weight:
332.37
UNSPSC Code:
12352005

Assay

≥95%

form

powder

mp

92-94 °C

storage temp.

−20°C

Application

Methyl (Z)-2-phenyl-3-(tosyloxy)acrylate is a versatile building block used for the preparation of α,β-disubstituted α,β-unsaturated esters through a variety of stereoretentive cross-couplings.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hidefumi Nakatsuji et al.
Organic letters, 11(19), 4258-4261 (2009-09-01)
An (E)- and (Z)-stereocomplementary preparative method for alpha,beta-disubstituted alpha,beta-unsaturated esters is performed via three general and robust reaction sequences: (i) Ti-Claisen condensation (formylation) of esters to give alpha-formyl esters (12 examples, 60-99%), (ii) (E)- and (Z)-stereocomplementary enol p-toluenesulfonylation (tosylation) using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service