Skip to Content
Merck
All Photos(3)

Key Documents

906751

Sigma-Aldrich

4-Vinylbenzyl(triphenyl)phosphonium chloride

≥98%

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€106.00
500 G
€620.00

€106.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€106.00
500 G
€620.00

About This Item

Empirical Formula (Hill Notation):
C27H24ClP
Molecular Weight:
414.91
UNSPSC Code:
12162002
NACRES:
NA.23

€106.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98%
98% (AgNO3, titration)

form

powder

contains

Contains 50 ppm of 4-tert-butylcatechol as stabilizer

color

white to light yellow

storage temp.

2-8°C

Application

The polymer of 4-Vinylbenzyl(triphenyl)phosphonium chloride exhibits UCST behaviour in the presence of sodium salts (NaCl, NaBr, and NaI) . In the presence of these salts, the polymer shows thermo responsiveness.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

How to manipulate the upper critical solution temperature (UCST).
Niskanen J, et al.
Polym. Chem., 8, 220-232 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service