Skip to Content
Merck
All Photos(2)

Key Documents

905186

Sigma-Aldrich

(9-Bromononyl)triphenylphosphonium bromide

Synonym(s):

Bromononane TPP mitochondrial tag, Mitochondria-targeting probe building block, Phosphonium lipocation tag

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H33Br2P
CAS Number:
Molecular Weight:
548.33
UNSPSC Code:
12352200

form

liquid

storage temp.

2-8°C

InChI

1S/C27H33BrP.BrH/c28-23-15-4-2-1-3-5-16-24-29(25-17-9-6-10-18-25,26-19-11-7-12-20-26)27-21-13-8-14-22-27;/h6-14,17-22H,1-5,15-16,23-24H2;1H/q+1;/p-1

InChI key

ACKJFSDDEOFXHT-UHFFFAOYSA-M

Application

Mitochondria perform several functions in the cell, and dysfunction has been implicated in various organ systems and disease states, including neurodegenerative disorders and cancers. The targeting of small molecules to the mitochondria can help to probe and better understand this relationship. This bromononane triphenylphosphine (TPP) tag is conjugated to small molecules to localize them to the mitochondria in cells where the TPP cations facilitate both cellular uptake and accumulation in the mitochondria.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francisco José Fueyo González et al.
Journal of medicinal chemistry, 60(4), 1509-1522 (2017-01-24)
We investigated a chemical strategy to boost the trypanocidal activity of 2,4-dihydroxybenzoic acid (2,4-DHBA)- and salicylhydroxamic acid (SHAM)-based trypanocides with triphenylphosphonium and quinolinium lipophilic cations (LC). Three series of LC conjugates were synthesized that were active in the submicromolar (5a-d
Michael P Murphy
Biochimica et biophysica acta, 1777(7-8), 1028-1031 (2008-04-29)
Mitochondrial function and dysfunction contributes to a range of important aspects of biomedical research. Consequently there is considerable interest in developing approaches to modify and report on mitochondria in cells and in vivo. One approach has been to target bioactive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service