Skip to Content
Merck
All Photos(1)

Key Documents

902373

Sigma-Aldrich

Glyceryl 1,3-dioctadecanoate-2-octanoate-1-13C

endotoxin tested, ≥99 atom % 13C, ≥99% (CP)

Synonym(s):

13C Labeled glyceryl 1,3-dioctadecanoate-2-octanoate, Glyceryl 1,3-distearate-3-ocatanoate-1-13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC46H90O6
Molecular Weight:
752.21
MDL number:
UNSPSC Code:
12352108

isotopic purity

≥99 atom % 13C

Assay

≥99% (CP)

form

solid

suitability

endotoxin tested

mass shift

M+1

SMILES string

CCCCCCCCCCCCCCCCC[13C](=O)OCC(CO[13C](=O)CCCCCCCCCCCCCCCCC)O[13C](=O)CCCCCCC

InChI

1S/C47H90O6/c1-4-7-10-13-15-17-19-21-23-25-27-29-31-34-36-39-45(48)51-42-44(53-47(50)41-38-33-12-9-6-3)43-52-46(49)40-37-35-32-30-28-26-24-22-20-18-16-14-11-8-5-2/h44H,4-43H2,1-3H3/i47+1

InChI key

ROSIALNPQYBSCH-RBMVYKJLSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

For R&D use only. Not for drug, household, or other uses. Any other use is the sole responsibility of the end user.




Endotoxin testing is done on the bulk form of the product before subdivision packaging. This bulk test does not guarantee that the subdivision or repackaged aliquot is endotoxin-free when it is received or used by the customer and does not imply suitability for any particular purpose. If the product must be endotoxin-free for the intended application, the product should be tested prior to actual use.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service