Skip to Content
Merck
All Photos(1)

Key Documents

900962

Sigma-Aldrich

N2300

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C54H72N2O4S2)n
UNSPSC Code:
12352200

description

Band gap: 1.68 eV

form

solid

mol wt

average Mn 25-50 kDa

color

blue to dark blue

solubility

anisole: soluble
chloroform: soluble
dichlorobenzene: soluble
toluene: soluble
xylene: soluble

Orbital energy

HOMO -5.58 eV 
LUMO -3.9 eV 

PDI

1.5‑3.5

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service