Skip to Content
Merck
All Photos(2)

Documents

900789

Sigma-Aldrich

Sodium 6-methylpyridine-2-sulfinate

≥95%

Synonym(s):

Willis 2-pyridylsulfinates Me

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6NNaO2S
Molecular Weight:
179.17
UNSPSC Code:
12352200

Assay

≥95%

form

powder

reaction suitability

reaction type: C-C Bond Formation

mp

314-319 °C

Application

This 2-pyridylsulfinate enables Suzuki-Miyaura cross coupling with aryl halides to access diverse pyridine derivatives as reported by the laboratory of Michael Willis.. No longer impeded by unstable 2-pyridyl boronates, the sulfinate alternatives provide Pd-catalyzed coupling routes to therapeutically valuable pyridine rings.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Description: Professor Willis and partners at Pfizer have shown with pyridine-2- sulfinates a palladium-catalyzed desulfinylative cross-coupling process can be realized.

Related Content

Research in the Willis group is focused on the development of new catalysts and reactions for synthetic chemistry. The group is particularly interested in addressing synthetic challenges that are applicable to the pharmaceutical and agrochemical industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service