Skip to Content
Merck
All Photos(1)

Key Documents

900162

Sigma-Aldrich

Ethyl pyruvate-1-13C

≥99 atom % 13C, ≥98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO13CO2CH2CH3
Molecular Weight:
117.11
UNSPSC Code:
51113400
PubChem Substance ID:

isotopic purity

≥99 atom % 13C

Assay

≥98% (CP)

form

liquid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+1

storage temp.

2-8°C

SMILES string

CCO[13C](C(C)=O)=O

InChI

1S/C5H8O3/c1-3-8-5(7)4(2)6/h3H2,1-2H3/i5+1

InChI key

XXRCUYVCPSWGCC-HOSYLAQJSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean-Michel Fernandes et al.
International journal of food sciences and nutrition, 72(1), 45-56 (2020-05-19)
Rice represents a primary source of carbohydrates in human nutrition. Upon its consumption, the released sugars are mostly absorbed, categorising rice as a high glycemic index food. Addition of ingredients is common practice when cooking rice, which may affect rice

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service