Skip to Content
Merck
All Photos(1)

Key Documents

795607

Sigma-Aldrich

4-Methoxybenzylphosphonic acid

Synonym(s):

MeOBPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11O4P
CAS Number:
Molecular Weight:
202.14
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

SMILES string

OP(CC1=CC=C(OC)C=C1)(O)=O

InChI

1S/C8H11O4P/c1-12-8-4-2-7(3-5-8)6-13(9,10)11/h2-5H,6H2,1H3,(H2,9,10,11)

InChI key

XUAVFSMXCWTYNM-UHFFFAOYSA-N

Application

Tuning of surface energy, wetting, and work function of metal oxides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marek Murias et al.
Bioorganic & medicinal chemistry, 12(21), 5571-5578 (2004-10-07)
Resveratrol (3,5,4'-trihydroxy-trans-stilbene) is found in grapes and various medical plants. Among cytotoxic, antifungal, antibacterial cardioprotective activity resveratrol also demonstrates non-selective cyclooxygenase-1 (COX-1) and cyclooxygenase-2 (COX-2) inhibition. In order to find more selective COX-2 inhibitors a series of methoxylated and hydroxylated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service