Skip to Content
Merck
All Photos(1)

Key Documents

793817

Sigma-Aldrich

5-Boc-2-oxa-5-azaspiro[3.4]octane

Synonym(s):

2-Oxa-5-azaspiro[3.4]octane-5-carboxylic acid tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO3
CAS Number:
Molecular Weight:
213.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

storage temp.

−20°C

SMILES string

CC(OC(N1CCCC21COC2)=O)(C)C

InChI

1S/C11H19NO3/c1-10(2,3)15-9(13)12-6-4-5-11(12)7-14-8-11/h4-8H2,1-3H3

InChI key

SDLALJACTHVVOO-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service