Skip to Content
Merck
All Photos(1)

Documents

78926

Sigma-Aldrich

(R)-(−)-1-Phenyl-2-propanol

≥97.0% (sum of enantiomers, GC)

Synonym(s):

(R)-(−)-α-Methylphenethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12O
CAS Number:
Molecular Weight:
136.19
Beilstein:
3195620
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Assay

≥97.0% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D −41±1°, c = 5.3% in benzene

optical purity

enantiomeric ratio: ≥99:1 (GC)

refractive index

n20/D 1.521

bp

95-97 °C/11 mmHg (lit.)

density

0.993 g/mL at 20 °C (lit.)

SMILES string

C[C@@H](O)Cc1ccccc1

InChI

1S/C9H12O/c1-8(10)7-9-5-3-2-4-6-9/h2-6,8,10H,7H2,1H3/t8-/m1/s1

InChI key

WYTRYIUQUDTGSX-MRVPVSSYSA-N

Other Notes

Chiral alcohol; it can be transformed to the chloride (SOCl2/pyridine) with inversion of configuration. The chloride undergoes AlCl3-catalyzed retentive Friedel-Crafts alkylation of benzene

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S. Masuda et al.
Bulletin of the Chemical Society of Japan, 56, 1089-1089 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service