Skip to Content
Merck
All Photos(1)

Key Documents

758361

Sigma-Aldrich

2-(4-Methyl-2-pyridyl)-1H-benzimidazole

99% (GC)

Synonym(s):

4-Methyl-2-(2-benzimidazolyl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11N3
CAS Number:
Molecular Weight:
209.25
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99% (GC)

form

solid

mp

225-230 °C

SMILES string

Cc1ccnc(c1)-c2nc3ccccc3[nH]2

InChI

1S/C13H11N3/c1-9-6-7-14-12(8-9)13-15-10-4-2-3-5-11(10)16-13/h2-8H,1H3,(H,15,16)

InChI key

MGBRKEZZOSQHQO-UHFFFAOYSA-N

General description

Such benzimidazole derivatives are usually associated with various kinds of pharmacokinetic and pharmacodynamic properties. Their heterocyclic structure possess wide range of antimicrobial properties.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benzimidazoles: A biologically active compounds
Salahuddin,Shaharyar M, Mazumdar A
Arabian Journal of Chemistry (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service