Skip to Content
Merck
All Photos(1)

Key Documents

756121

Sigma-Aldrich

Boron trifluoride diethyl-d10 etherate

98 atom % D, 95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4D10BF3O
Molecular Weight:
151.99
UNSPSC Code:
12352005

isotopic purity

98 atom % D

Assay

95% (CP)

form

liquid

bp

126-129 °C (lit.)

mp

-58 °C (lit.)

density

1.23 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[D3C]C([2H])([2H])O(C([2H])([2H])C([2H])([2H])[2H])B(F)(F)F

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT RE 1 Inhalation

Target Organs

Kidney

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

118.4 °F

Flash Point(C)

48 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service