Skip to Content
Merck
All Photos(1)

Key Documents

754536

Sigma-Aldrich

9-[2-(Diisopropylphosphino)phenyl]-9H-carbazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26NP
CAS Number:
Molecular Weight:
359.44
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

145-149 °C

functional group

phosphine

SMILES string

CC(C)P(C(C)C)c1ccccc1-n2c3ccccc3c4ccccc24

InChI

1S/C24H26NP/c1-17(2)26(18(3)4)24-16-10-9-15-23(24)25-21-13-7-5-11-19(21)20-12-6-8-14-22(20)25/h5-18H,1-4H3

InChI key

LVZFAAVQZRSWHJ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service