Skip to Content
Merck
All Photos(1)

Key Documents

752320

Sigma-Aldrich

S-(3-Azidopropyl)thioacetate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9N3OS
Molecular Weight:
159.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.506

density

1.139 g/mL at 25 °C

functional group

azide
thioester

storage temp.

2-8°C

SMILES string

CC(=O)SCCCN=[N+]=[N-]

InChI

1S/C5H9N3OS/c1-5(9)10-4-2-3-7-8-6/h2-4H2,1H3

InChI key

UHSOFTIGNARKFK-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.9 °F

Flash Point(C)

109.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service