Skip to Content
Merck
All Photos(1)

Key Documents

749818

Sigma-Aldrich

Indium tin oxide coated PET

surface resistivity 300 Ω/sq, L × W × thickness 1 ft × 1 ft × 7 mil, sheet

Synonym(s):

ITO-PET, Polyethylene terephthalate film, ITO coated

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
In2O3 · (SnO2)x
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

sheet

surface resistivity

300 Ω/sq

L × W × thickness

1 ft × 1 ft × 7 mil

SMILES string

O=[In]O[In]=O.O=[Sn]=O

InChI

1S/2In.5O.Sn

InChI key

LNNWKAUHKIHCKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indium tin oxide coated polyethylene terephthalate (PET) film is optically transparent and electrically conductive.[1]The ITO coating is protected by a thin, transparent film, that can be easily peeled off.

Application

ITO-PET may be used in fabricating flexible OLEDs.[2] ITO/PET may be used as a reference anode in conventional flexible polymer based organic solar cells (OSCs).[3]

Features and Benefits

The polyester film products comprise sputtered ITO, and exhibit transmittance and resistance characteristics similar to those of the glass substrates.
These polyester film products comprise of sputtered ITO, and exhibit transmittance and resistance characteristics similar to those of glass substrates.

Physical properties

Thickness of ITO coating is 1000 Å

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modular mass spectrometric tool for analysis of composition and phosphorylation of protein complexes
Blethrow JD, et al.
PLoS ONE, 2(4), e358-e358 (2007)
High-Performance Organic Light-Emitting Diodes Using ITO Anodes Grown on Plastic by Room-Temperature Ion-Assisted Deposition
Yang Y, et al.
Advanced Materials, 16(4), 321-324 (2004)
Efficient and Flexible ITO-Free Organic Solar Cells Using Highly Conductive Polymer Anodes.
Na SI, et al.
Advanced Materials, 20(21), 4061-4067 (2008)

Articles

A transparent conductive electrode (TCE) is an essential component of various optoelectronic devices such as solar cells, liquid-crystal displays (LCD), light-emitting diodes (LED), and touch screens.

Progress in solution-processed functional materials leads to thin-film optoelectronic devices for industrial and consumer electronics.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Questions

1–4 of 4 Questions  
  1. How do I determine which side of this product has the Indium Tin Oxide (ITO) coating?

    1 answer
    1. The ITO side of the PET sheet has a blue protective film on it to prevent the ITO from scratching.  Once this film is removed, you can also determine the coated side by testing for ITO resistance using either a Multimeter or a Four Point Probe.

      Helpful?

  2. Why isn't the Indium tin oxide (ITO) coated PET showing resistance?

    1 answer
    1. The Indium tin oxide (ITO) coated PET is protected by a thin, transparent film that can be easily peeled off.  This film will not show resistance, so it needs to be removed prior to testing the electrical properties.

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. What is the light transmittance of Products 749737, 749753, 749788, and 749818, Indium tin oxide coated PET, at 550 nm?

    1 answer
    1. Our supplier has provided the transmittance values for the materials at 550 nm. These values are as follows:749737 Indium tin oxide coated PET 100 Ohms/sq- greater than 77.0% Transmittance at 550 nm749753 Indium tin oxide coated PET 200 Ohms/sq- greater than 82.5% Transmittance at 550 nm749788 Indium tin oxide coated PET 250 Ohms/sq- greater than 83.0% Transmittance at 550 nm749818 Indium tin oxide coated PET 300 Ohms/sq- greater than 84.0% Transmittance at 550 nm.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service