Skip to Content
Merck
All Photos(1)

Key Documents

746983

Sigma-Aldrich

Poly(styrene)-block-poly(acrylic acid)

average Mn 38,000, 20 wt. % PAA, PDI ≤1.3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N(C8H8)m(C3H4O2)nH
UNSPSC Code:
12352100
NACRES:
NA.23

form

powder

mol wt

Mn 27,000-33,000 (polystyrene)
Mn 34,000-42,000 (total)
Mn 7,000-9,000 (poly(acrylic acid))
average Mn 38,000

PDI

≤1.3

Looking for similar products? Visit Product Comparison Guide

Application

Polystyrene-block-polyacrylic acid is a diblock copolymer used for making polymeric micelles/vesicles (polymersomes) and other encapsulation applications[1],[2]. This PS-block-PAA copolymer is 20 wt. % AA; and should form micelles in water in the 30~50 nm range. The polystyrene degree of polymerization (DP) is 275 and the polyacrylic acid DP is 120.

Legal Information

Sold for research purposes only and subject to field restriction: see www.sigmaaldrich.com/raftlicense.  Patents: 7,714,075; 7,250,479; 7,666,962; 6,642,318; 6,747,111; W2010/8356.

Sold for research purposes only.  Not for use in humans.  Before opening or using this product, please read the following terms and conditions.  Use of this product shall constitute acknowledgement and acceptance of these terms and conditions.
Use of this product may be covered by one or more of the following US patents and corresponding claims outside the US: 7,714,075; 7,250,479; 7,666,962; 6,642,318; 6,747,111, and pending applications (W2010/8356).  For details contact Aldrich Chemical Company, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dennis E Discher et al.
Science (New York, N.Y.), 297(5583), 967-973 (2002-08-10)
Vesicles are microscopic sacs that enclose a volume with a molecularly thin membrane. The membranes are generally self-directed assemblies of amphiphilic molecules with a dual hydrophilic-hydrophobic character. Biological amphiphiles form vesicles central to cell function and are principally lipids of
Kaoutar Moumile et al.
Journal of clinical microbiology, 42(2), 923-924 (2004-02-10)
We describe a case of primary purulent culture-negative pericarditis caused by Neisseria meningitidis serogroup C occurring in an 8-month-old previously healthy boy, which was detected in pericardial fluid by broad-spectrum PCR amplification.

Articles

Reversible addition–fragmentation chain transfer (RAFT) polymerization is rapidly moving to the forefront in construction of drug and gene delivery vehicles.

The development of drugs that target specific locations within the human body remains one of the greatest challenges in biomedicine today.

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

Over the past two decades, the rapid advance of controlled living polymerization (CLP) techniques.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service