Skip to Content
Merck
All Photos(1)

Key Documents

745308

Sigma-Aldrich

1-(Azidomethyl)-4-fluorobenzene solution

~0.5 M in dichloromethane, ≥97.0% (HPLC)

Synonym(s):

p-Fluorobenzyl azide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6FN3
CAS Number:
Molecular Weight:
151.14
Beilstein:
8403292
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
To inquire about this product 745308, please contact your local Merck office or dealer. Contact Technical Service

Assay

≥97.0% (HPLC)

form

solution

concentration

~0.5 M in dichloromethane

impurities

≤2.0% water

storage temp.

2-8°C

SMILES string

Fc1ccc(CN=[N+]=[N-])cc1

InChI

1S/C7H6FN3/c8-7-3-1-6(2-4-7)5-10-11-9/h1-4H,5H2

InChI key

UEQMFQIPTDUPPJ-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 2 Dermal - STOT SE 3

Target Organs

Central nervous system, Lungs,Nervous system,Cardio-vascular system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave-assisted benzyne-click chemistry: preparation of 1H-benzo[d][1,2,3]triazoles
Ankatia, H. and Biehl, E.
Tetrahedron Letters, 50, 4677-4682 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service