Skip to Content
Merck
All Photos(1)

Key Documents

739766

Sigma-Aldrich

Methyl 2-iodo-5-methylbenzoate

95%

Synonym(s):

2-Iodo-5-methylbenzoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9IO2
CAS Number:
Molecular Weight:
276.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.596

density

1.674 g/mL at 25 °C

functional group

ester
iodo

storage temp.

2-8°C

SMILES string

COC(=O)c1cc(C)ccc1I

InChI

1S/C9H9IO2/c1-6-3-4-8(10)7(5-6)9(11)12-2/h3-5H,1-2H3

InChI key

DHYMLHOXGMUIDZ-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service