Skip to Content
Merck
All Photos(2)

Key Documents

733512

Sigma-Aldrich

Tin telluride

greener alternative

pieces, 99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
SnTe
CAS Number:
Molecular Weight:
246.31
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

Assay

99.999% trace metals basis

form

pieces

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

780.0 °C (lit.)

greener alternative category

SMILES string

[SnH2]=[Te]

InChI

1S/Sn.Te

InChI key

WYUZTTNXJUJWQQ-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Tin Telluride (SnTe) is a IV-VI semiconductor with an energy band gap of 0.18 eV at 300K. It can be used as a thermoelectric material, which can be used in the conversion of heat into electricity. It can be doped with indium and calcium to achieve a thermoelectric figure of merit, ZT of ~1.65 at 840 K and a power factor of ~47 μW cm-1 K-2.[1][2][3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced thermoelectric performance of bulk tin telluride: synergistic effect of calcium and indium co-doping
Bhat DK and Shenoy US
Chemosphere, 4(13), 12-18 (2018)
Volykhov, A. A.; Yashina, L. V.; Tamm, M. E.; Ryzhenkov, A. V.
Inorganic Materials, 45, 968-968 (2009)
Hoang, Khang; Mahanti, S. D.; Kanatzidis, Mercouri G.
Physica B: Condensed Matter, 81, 115106/1-115106/1 (2010)
Simple routes to synthesis and characterization of nanosized tin telluride compounds
Salavati-Niasari M, et al.
Applied Surface Science, 257(3), 781-785 (2010)
High thermoelectric performance of co-doped tin telluride due to synergistic effect of magnesium and indium
Bhat DK and Shenoy US
The Journal of Physical Chemistry C, 121(13), 7123-7130 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service