Skip to Content
Merck
All Photos(1)

Documents

727768

Sigma-Aldrich

H-Rink amide ChemMatrix® resin

35-100 mesh particle size (wet sieved)

Synonym(s):

4-[(2,4-Dimethoxyphenyl)(amino)methyl]phenoxyacetic acid, polymer-bound to ChemMatrix® resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

product line

ChemMatrix®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

0.40-0.60 mmol/g loading

particle size

35-100 mesh (wet sieved)

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

SMILES string

O=C(COC1=CC=C(C(C2=CC=C(OC)C=C2OC)NC(OCC3C4=C(C=CC=C4)C5=C3C=CC=C5)=O)C=C1)NC[*]

General description

H-Rink amide ChemMatrix® resin is a poly(ethylene glycol) (PEG)-based resin with high stability due to the primary ether bonds of poly(ethylene glycol).

Application

H-Rink amide ChemMatrix® resin can be used for solid phase peptide synthesis (SPPS).

Legal Information

ChemMatrix is a registered trademark of Matrix Innovation, Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Green solid-phase peptide synthesis 2. 2-Methyltetrahydrofuran and ethyl acetate for solid-phase peptide synthesis under green conditions.
Jad YE, et al.
ACS sustainable chemistry & engineering, 4(12), 6809-6814 (2016)
ChemMatrix for complex peptides and combinatorial chemistry.
Garcia-Ramos Y, et al.
Journal of Peptide Science, 16(12), 675-678 (2010)
ChemMatrix, a poly (ethylene glycol)-based support for the solid-phase synthesis of complex peptides
Garcia-Martin F, et al.
Journal of Combinatorial Chemistry, 8(2), 213-220 (2006)

Articles

ChemMatrix® is a proprietary, 100% PEG (polyethylene glycol) based resin from PCAS BioMatrix.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service