Skip to Content
Merck
All Photos(1)

Key Documents

726877

Sigma-Aldrich

(S)-(−)-1-(2-Naphthyl)ethylamine

ChiPros®, produced by BASF, ≥98.5%

Synonym(s):

(S)-(−)-α-Methyl-2-naphthalenemethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13N
CAS Number:
Molecular Weight:
171.24
Beilstein:
3195949
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

produced by BASF

Assay

≥98.5%

form

powder

optical purity

enantiomeric excess: ≥98.5%

SMILES string

C[C@H](N)c1ccc2ccccc2c1

InChI

1S/C12H13N/c1-9(13)11-7-6-10-4-2-3-5-12(10)8-11/h2-9H,13H2,1H3/t9-/m0/s1

InChI key

KHSYYLCXQKCYQX-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service