Skip to Content
Merck
All Photos(1)

Documents

726869

Sigma-Aldrich

(S)-(−)-1-(4-Bromophenyl)ethylamine

ChiPros®, produced by BASF, 99%

Synonym(s):

(S)-(−)-4-Bromo-α-methylbenzylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10BrN
CAS Number:
Molecular Weight:
200.08
Beilstein:
5729988
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

produced by BASF

Assay

≥98.5% (GC)
99%

optical purity

enantiomeric excess: ≥98.5%

density

1.390 g/mL at 20 °C (lit.)

SMILES string

C[C@H](N)c1ccc(Br)cc1

InChI

1S/C8H10BrN/c1-6(10)7-2-4-8(9)5-3-7/h2-6H,10H2,1H3/t6-/m0/s1

InChI key

SOZMSEPDYJGBEK-LURJTMIESA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service