Skip to Content
Merck
All Photos(1)

Key Documents

717460

Sigma-Aldrich

Chloro[tri(o-tolyl)phosphine]gold(I)

95%

Synonym(s):

[Tri(2-methylphenyl)phosphine]gold(I) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21AuClP
CAS Number:
Molecular Weight:
536.78
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

95%

form

solid

reaction suitability

core: gold
reagent type: catalyst

SMILES string

Cl[Au].Cc1ccccc1P(c2ccccc2C)c3ccccc3C

InChI

1S/C21H21P.Au.ClH/c1-16-10-4-7-13-19(16)22(20-14-8-5-11-17(20)2)21-15-9-6-12-18(21)3;;/h4-15H,1-3H3;;1H/q;+1;/p-1

InChI key

VECXLHSTZNCQLD-UHFFFAOYSA-M

Application

Catalyst for:
  • Alkylation of silyl enol ethers with orthoalkynylbenzoic acid esters
  • Cycloisomerization
  • Cyclization of 1,6-enynes
  • Tandem [3,3]-sigmatropic rearrangement-cycloisomerization cascade
  • Cycloaddition of enyne derivatives with aryl alkynes or alkenes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service