Skip to Content
Merck
All Photos(1)

Documents

711098

Sigma-Aldrich

Potassium pyrimidine-5-trifluoroborate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3BF3KN2
CAS Number:
Molecular Weight:
185.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

99-103 °C

SMILES string

[K+].F[B-](F)(F)c1cncnc1

InChI

1S/C4H3BF3N2.K/c6-5(7,8)4-1-9-3-10-2-4;/h1-3H;/q-1;+1

InChI key

FLVBHQQPIJARSC-UHFFFAOYSA-N

Application

Organotrifluoroborate involved in C-O activation and cross-coupling reactions

Organotrifluoroborates as versatile and stable boronic acid surrogates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Bench-stable Potassium Organotrifluoroborates enable diverse C-C bond formation reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service